Product Details

SNP ID
rs199929675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20031739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTGCCACACCTATGGAATCAC[A/G]GGGATACTTTTATAGGTTTTCCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR139 PubMed Links

Gene Details

Gene
GPR139
Gene Name
G protein-coupled receptor 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002911.3 1220 Missense Mutation CCG,CTG P353L NP_001002911.1
NM_001318483.1 1220 Missense Mutation CCG,CTG P260L NP_001305412.1

View Full Product Details