Product Details

SNP ID
rs200191944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30570329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCACGCTAATAGCTGGGTCAG[C/G]GTTGCGTTCCACCAGCACTTCAGGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF688 PubMed Links

Gene Details

Gene
ZNF688
Gene Name
zinc finger protein 688
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024683.1 508 Missense Mutation CCT,GCT P126A NP_001019854.1
NM_145271.3 508 Missense Mutation CCT,GCT P140A NP_660314.1
XM_005255139.2 508 Missense Mutation CCT,GCT P63A XP_005255196.1
XM_005255140.2 508 Missense Mutation CCT,GCT P63A XP_005255197.1
Gene
ZNF785
Gene Name
zinc finger protein 785
There are no transcripts associated with this gene.

View Full Product Details