Product Details

SNP ID
rs199475705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5501463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATCATCTTTGTAGAAAACTAGA[A/T]ATTGAGAAAACAGTGTAGGCAAAAT
Phenotype
MIM: 606636
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC728392 PubMed Links

Gene Details

Gene
LOC728392
Gene Name
uncharacterized LOC728392
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162371.2 5034 Intron NP_001155843.1
Gene
NLRP1
Gene Name
NLR family pyrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033053.2 5034 UTR 3 NP_001028225.1
NM_014922.4 5034 Intron NP_055737.1
NM_033004.3 5034 Intron NP_127497.1
NM_033006.3 5034 Intron NP_127499.1
NM_033007.3 5034 Intron NP_127500.1

View Full Product Details