Product Details

SNP ID
rs199769574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9022712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCTTCCTCACCGACCTCAACAA[C/T]CCGCACAACCTGACGTGCTGGCAGT
Phenotype
MIM: 601614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTN1 PubMed Links

Gene Details

Gene
NTN1
Gene Name
netrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004822.2 546 Silent Mutation AAC,AAT N113N NP_004813.2
XM_006721595.3 546 Silent Mutation AAC,AAT N113N XP_006721658.1

View Full Product Details