Product Details

SNP ID
rs200184901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48622974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAAACCAGATTTTGACTTGTC[C/T]CTCACTCAGATTGAGGAGTCTGGCC
Phenotype
MIM: 142963 MIM: 142964 MIM: 608632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB8 PubMed Links

Gene Details

Gene
HOXB8
Gene Name
homeobox B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024016.3 883 Intron NP_076921.1
XM_005257286.2 883 Intron XP_005257343.1
XM_017024564.1 883 Intron XP_016880053.1
Gene
HOXB9
Gene Name
homeobox B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024017.4 883 Missense Mutation AGA,GGA R227G NP_076922.1
Gene
MIR196A1
Gene Name
microRNA 196a-1
There are no transcripts associated with this gene.

View Full Product Details