Product Details

SNP ID
rs200025811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGGGAATTTCCTTATGGATGA[C/T]AACCATCTGCCTGAAACTTTCCTCC
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 989 Missense Mutation ATC,GTC I282V NP_001307881.1
NM_001320953.1 989 Missense Mutation ATC,GTC I264V NP_001307882.1
NM_001320954.1 989 Missense Mutation ATC,GTC I264V NP_001307883.1
NM_001320955.1 989 Missense Mutation ATC,GTC I263V NP_001307884.1
NM_014519.3 989 Missense Mutation ATC,GTC I291V NP_055334.2
XM_011524006.2 989 Missense Mutation ATC,GTC I243V XP_011522308.1
XM_017025021.1 989 Missense Mutation ATC,GTC I291V XP_016880510.1
XM_017025022.1 989 Missense Mutation ATC,GTC I215V XP_016880511.1
XM_017025023.1 989 Missense Mutation ATC,GTC I206V XP_016880512.1

View Full Product Details