Product Details

SNP ID
rs201810621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38320686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGAATCTTTAGAGCCCTCTCA[C/T]GTTGTTCAAGTTCGCTGTTCAAGTA
Phenotype
MIM: 614515 MIM: 611850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR179 PubMed Links

Gene Details

Gene
GPR179
Gene Name
G protein-coupled receptor 179
There are no transcripts associated with this gene.

Gene
MRPL45
Gene Name
mitochondrial ribosomal protein L45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278279.2 740 Intron NP_001265208.2
NM_032351.5 740 Silent Mutation CAC,CAT H193H NP_115727.5

View Full Product Details