Product Details

SNP ID
rs201314485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18969129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCTCTCCACCATCCTCACGCT[C/T]GGCATCACAGCCCTGTACACCATCG
Phenotype
MIM: 615886
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM83G PubMed Links

Gene Details

Gene
FAM83G
Gene Name
family with sequence similarity 83 member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039999.2 4953 Intron NP_001035088.2
XM_017024953.1 4953 UTR 3 XP_016880442.1
Gene
SLC5A10
Gene Name
solute carrier family 5 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042450.2 4953 Silent Mutation CTC,CTT L177L NP_001035915.1
NM_001270648.1 4953 Silent Mutation CTC,CTT L177L NP_001257577.1
NM_001270649.1 4953 Silent Mutation CTC,CTT L177L NP_001257578.1
NM_001282417.1 4953 Silent Mutation CTC,CTT L121L NP_001269346.1
NM_152351.4 4953 Silent Mutation CTC,CTT L177L NP_689564.3
XM_011523652.1 4953 Silent Mutation CTC,CTT L219L XP_011521954.1
XM_011523653.2 4953 Silent Mutation CTC,CTT L219L XP_011521955.1
XM_017024191.1 4953 Silent Mutation CTC,CTT L219L XP_016879680.1

View Full Product Details