Product Details
- SNP ID
-
rs199719131
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19711368 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTATATTCTTAGACTTTCTCTCCC[A/C]TATGAGTCCTTTCATGCAGTCGAAA
- Phenotype
-
MIM: 194556
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF14
PubMed Links
Gene Details
- Gene
- ZNF14
- Gene Name
- zinc finger protein 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021030.2 |
2067 |
Missense Mutation |
AGG,ATG |
R638M |
NP_066358.2 |
View Full Product Details