Product Details

SNP ID
rs199553590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41425587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCGCCCTCCCCAATGAGAGTC[A/C]GCACTGGAGCCTTGGGTCTTGCAGT
Phenotype
MIM: 615357 MIM: 608348
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5SL PubMed Links

Gene Details

Gene
ATP5SL
Gene Name
ATP5S like
There are no transcripts associated with this gene.

Gene
B3GNT8
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
There are no transcripts associated with this gene.

Gene
BCKDHA
Gene Name
branched chain keto acid dehydrogenase E1, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000709.3 Intron NP_000700.1
NM_001164783.1 Intron NP_001158255.1

View Full Product Details