Product Details

SNP ID
rs199871361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56003953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATGCTCTATTCCACAGTTTGA[A/C]ATCGAGAATGCCAACGTGGAATGTC
Phenotype
MIM: 609658
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NLRP5 PubMed Links

Gene Details

Gene
NLRP5
Gene Name
NLR family pyrin domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153447.4 300 Missense Mutation GAA,GAC E100D NP_703148.4

View Full Product Details