Product Details

SNP ID
rs200020270
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56664233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTGTGTGGATGCGCCGGTGCTC[C/G]GCCAGAGAGGCGCTCTGGCTGAAGA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SMIM17 PubMed Links

Gene Details

Gene
SMIM17
Gene Name
small integral membrane protein 17
There are no transcripts associated with this gene.

Gene
ZNF835
Gene Name
zinc finger protein 835
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005850.2 1228 Silent Mutation GCC,GCG A322A NP_001005850.2
XM_005259382.2 1228 Silent Mutation GCC,GCG A322A XP_005259439.1
XM_005259383.3 1228 Silent Mutation GCC,GCG A322A XP_005259440.1

View Full Product Details