Product Details

SNP ID
rs200892720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56088066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCACTCGGGGCACCGCCCGCCC[C/G]CGGCCTCGTCGTCGTCGTCCTCCTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928886 PubMed Links

Gene Details

Gene
LOC101928886
Gene Name
uncharacterized LOC101928886
There are no transcripts associated with this gene.

Gene
ZNF787
Gene Name
zinc finger protein 787
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002836.3 1318 Missense Mutation GCG,GGG A369G NP_001002836.2
XM_011526445.2 1318 Missense Mutation GCG,GGG A369G XP_011524747.1

View Full Product Details