Product Details

SNP ID
rs200589534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52155003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACTAATGAAAGATTTGCCACACT[C/T]ATTACATTTATAAGGTTTTTCTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF836 PubMed Links

Gene Details

Gene
ZNF836
Gene Name
zinc finger protein 836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102657.1 3089 Missense Mutation AAG,GAG K894E NP_001096127.1
XM_011526558.2 3089 Missense Mutation AAG,GAG K894E XP_011524860.1
XM_011526559.2 3089 Missense Mutation AAG,GAG K894E XP_011524861.1
XM_017026389.1 3089 Intron XP_016881878.1

View Full Product Details