Product Details

SNP ID
rs201870668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:464062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCATGGTGAAGGCAGGCAGGC[A/G]CTGGCGGTAGGTGTTTGCGTCCGGG
Phenotype
MIM: 605217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ODF3L2 PubMed Links

Gene Details

Gene
ODF3L2
Gene Name
outer dense fiber of sperm tails 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182577.2 458 Missense Mutation CGC,TGC R218C NP_872383.1
XM_005259545.4 458 Missense Mutation CGC,TGC R110C XP_005259602.1
XM_017026671.1 458 Missense Mutation CGC,TGC R110C XP_016882160.1
Gene
SHC2
Gene Name
SHC adaptor protein 2
There are no transcripts associated with this gene.

View Full Product Details