Product Details

SNP ID
rs200563568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39391409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTCGTAACGCACTTCCGGCGG[C/T]CTACGCGAGGAAGATGGCTGCATCC
Phenotype
MIM: 612914 MIM: 610506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED29 PubMed Links

Gene Details

Gene
MED29
Gene Name
mediator complex subunit 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317770.1 107 Missense Mutation GCC,GTC A17V NP_001304699.1
NM_017592.2 107 Missense Mutation GCC,GTC A17V NP_060062.1
Gene
PAF1
Gene Name
PAF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256826.1 107 Intron NP_001243755.1
NM_019088.3 107 Intron NP_061961.2
Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303614.1 107 Intron NP_001290543.1
NM_018028.3 107 Intron NP_060498.2
XM_011527057.2 107 Intron XP_011525359.1
XM_011527060.2 107 Intron XP_011525362.1
XM_011527061.2 107 Intron XP_011525363.1
XM_011527062.2 107 Intron XP_011525364.1
XM_011527063.2 107 Intron XP_011525365.1
XM_011527064.2 107 Intron XP_011525366.1
XM_017026918.1 107 Intron XP_016882407.1
XM_017026919.1 107 Intron XP_016882408.1
XM_017026920.1 107 Intron XP_016882409.1
XM_017026921.1 107 Intron XP_016882410.1
XM_017026922.1 107 Intron XP_016882411.1
XM_017026923.1 107 Intron XP_016882412.1

View Full Product Details