Product Details

SNP ID
rs201225013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54769872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATGTCGCTCTTGGTCGTCAGC[A/G]TGGCGTGTGTTGGTGAGTCCTGGAA
Phenotype
MIM: 604936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIR2DL1 PubMed Links

Gene Details

Gene
KIR2DL1
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014218.2 665 Missense Mutation ATG,GTG M8V NP_055033.2
XM_011526939.2 665 Intron XP_011525241.1
XM_017026782.1 665 Intron XP_016882271.1
XM_017026783.1 665 Missense Mutation ATG,GTG M8V XP_016882272.1
Gene
LOC101928804
Gene Name
uncharacterized LOC101928804
There are no transcripts associated with this gene.

View Full Product Details