Product Details

SNP ID
rs201763306
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:19284005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGTCACTGGTTACCTGCAGCTG[C/T]GAGTGGTGGTACATCCACCGGAGGG
Phenotype
MIM: 608477 MIM: 608478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR7A3 PubMed Links

Gene Details

Gene
AKR7A3
Gene Name
aldo-keto reductase family 7 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012067.2 636 Silent Mutation TCA,TCG S275S NP_036199.2
XM_011541046.2 636 Silent Mutation TCA,TCG S275S XP_011539348.1
XM_017000714.1 636 Silent Mutation TCA,TCG S204S XP_016856203.1
Gene
AKR7L
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
LOC100506730
Gene Name
uncharacterized LOC100506730
There are no transcripts associated with this gene.

View Full Product Details