Product Details

SNP ID
rs201191528
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:197086873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTATAGAAGAATTCTTCTTTT[G/T]CTTGTAAAGTATTCTTTCAGTATTC
Phenotype
MIM: 605481
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10518 Missense Mutation AAA,CAA K1836Q NP_001193775.1
NM_018136.4 10518 Missense Mutation AAA,CAA K3421Q NP_060606.3

View Full Product Details