Product Details

SNP ID
rs201612591
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:196982916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGATTTTCCAAAAATACACCA[A/T]GGATTTCTGTATGATGAAGAAGATT
Phenotype
MIM: 608593
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 218 Missense Mutation CAA,CAT Q30H NP_110414.1
XM_011510020.2 218 Missense Mutation CAA,CAT Q33H XP_011508322.1

View Full Product Details