Product Details

SNP ID
rs201158336
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:74199713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGACATAGTTGCAATAACAGC[A/G]GCTGATGAAAAACAGGAACTTGCTT
Phenotype
MIM: 603609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FPGT PubMed Links

Gene Details

Gene
FPGT
Gene Name
fucose-1-phosphate guanylyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199328.2 199 Silent Mutation GCA,GCG A57A NP_001186257.2
NM_001199329.2 199 Silent Mutation GCA,GCG A57A NP_001186258.2
NM_003838.4 199 Silent Mutation GCA,GCG A57A NP_003829.3
Gene
FPGT-TNNI3K
Gene Name
FPGT-TNNI3K readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112808.2 199 Silent Mutation GCA,GCG A57A NP_001106279.2
NM_001199327.1 199 Silent Mutation GCA,GCG A57A NP_001186256.2
Gene
LRRIQ3
Gene Name
leucine rich repeats and IQ motif containing 3
There are no transcripts associated with this gene.

View Full Product Details