Product Details

SNP ID
rs201053260
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:6206573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCGGGCCCCTGGAGGGCCCGA[A/G]CTCCCTGGATGCCCCGAGCATCCAC
Phenotype
MIM: 616923 MIM: 180474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724450 PubMed Links

Gene Details

Gene
LOC102724450
Gene Name
uncharacterized LOC102724450
There are no transcripts associated with this gene.

Gene
RNF207
Gene Name
ring finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207396.2 212 Missense Mutation AAC,AGC N13S NP_997279.2
XM_011541439.2 212 Silent Mutation GAA,GAG E105E XP_011539741.1
XM_017001259.1 212 Missense Mutation AAC,AGC N13S XP_016856748.1
Gene
RPL22
Gene Name
ribosomal protein L22
There are no transcripts associated with this gene.

View Full Product Details