Product Details

SNP ID
rs201067493
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:100085550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGCTTACCTGAATTACTAAAT[A/C]GGTTCTGGCTGAGTCCGCGTAAAGG
Phenotype
MIM: 609321
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFSD14A PubMed Links

Gene Details

Gene
MFSD14A
Gene Name
major facilitator superfamily domain containing 14A
There are no transcripts associated with this gene.

Gene
SASS6
Gene Name
SAS-6 centriolar assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304829.1 2054 Missense Mutation CGA,CTA R451L NP_001291758.1
NM_194292.2 2054 Missense Mutation CGA,CTA R618L NP_919268.1
XM_017000486.1 2054 Missense Mutation CGA,CTA R591L XP_016855975.1

View Full Product Details