Product Details
- SNP ID
-
rs199997424
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:12859983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTCAATTATCTAACGCCAATTA[A/C]ATATCTCAGAAAGTCATTGAAAATA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF2
PubMed Links
Gene Details
- Gene
- PRAMEF2
- Gene Name
- PRAME family member 2
View Full Product Details