Product Details

SNP ID
rs199987519
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:200874084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGACAGTTCCGTGTTCCGTTG[A/C]CGGGCCCAGGCCGCGAACACTGCCT
Phenotype
MIM: 602174
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 1130 Silent Mutation TGA,TGC *349C NP_005289.2

View Full Product Details