Product Details

SNP ID
rs200739616
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:200873942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGGCCTTCGTCAACAGCTGCG[C/G]CAACCCGCTCATCTACCTCCTGCTG
Phenotype
MIM: 602174
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 988 Missense Mutation GCC,GGC A302G NP_005289.2

View Full Product Details