Product Details

SNP ID
rs200215834
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:165827851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGAACCATGGCCGGGGACAGCGA[A/G]CAGACCCTGCAGAACCACCAGCAGC
Phenotype
MIM: 609329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UCK2 PubMed Links

Gene Details

Gene
UCK2
Gene Name
uridine-cytidine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012474.4 357 Silent Mutation GAA,GAG E6E NP_036606.2
XM_011509970.2 357 Intron XP_011508272.2
XM_017002236.1 357 UTR 5 XP_016857725.1

View Full Product Details