Product Details

SNP ID
rs202240787
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:6207401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACACCAGACGGTGCTGAAGGGT[C/T]CCAGCGGGCTCCCGCCGGTGGACCG
Phenotype
MIM: 616923 MIM: 180474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724450 PubMed Links

Gene Details

Gene
LOC102724450
Gene Name
uncharacterized LOC102724450
There are no transcripts associated with this gene.

Gene
RNF207
Gene Name
ring finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207396.2 388 Missense Mutation CCC,TCC P72S NP_997279.2
XM_011541439.2 388 Intron XP_011539741.1
XM_017001259.1 388 Missense Mutation CCC,TCC P72S XP_016856748.1
Gene
RPL22
Gene Name
ribosomal protein L22
There are no transcripts associated with this gene.

View Full Product Details