Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281437.1 | 10961 | Intron | NP_001268366.1 | ||
NM_001281438.1 | 10961 | Intron | NP_001268367.1 | ||
NM_007002.3 | 10961 | Intron | NP_008933.2 | ||
NM_175573.2 | 10961 | Intron | NP_783163.1 | ||
XM_005260257.1 | 10961 | Intron | XP_005260314.1 | ||
XM_011528503.1 | 10961 | Intron | XP_011526805.1 | ||
XM_017027602.1 | 10961 | Intron | XP_016883091.1 | ||
XM_017027603.1 | 10961 | Intron | XP_016883092.1 | ||
XM_017027604.1 | 10961 | Intron | XP_016883093.1 | ||
XM_017027605.1 | 10961 | Intron | XP_016883094.1 | ||
XM_017027606.1 | 10961 | Intron | XP_016883095.1 | ||
XM_017027607.1 | 10961 | Intron | XP_016883096.1 | ||
XM_017027608.1 | 10961 | Intron | XP_016883097.1 | ||
XM_017027609.1 | 10961 | Intron | XP_016883098.1 | ||
XM_017027610.1 | 10961 | Intron | XP_016883099.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005560.4 | 10961 | Missense Mutation | CGG,TGG | R3672W | NP_005551.3 |
XM_006723796.3 | 10961 | Missense Mutation | CGG,TGG | R3677W | XP_006723859.1 |
XM_006723798.3 | 10961 | Intron | XP_006723861.1 | ||
XM_011528818.2 | 10961 | Missense Mutation | CGG,TGG | R3631W | XP_011527120.1 |
XM_011528819.2 | 10961 | Missense Mutation | CGG,TGG | R3586W | XP_011527121.1 |