Product Details

SNP ID
rs200785757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34550001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAAGATGAGATTCTTCAGTTC[C/T]CCATGTGGAAAAGCAGCTGTGGACC
Phenotype
MIM: 607167 MIM: 601242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNLRB1 PubMed Links

Gene Details

Gene
DYNLRB1
Gene Name
dynein light chain roadblock-type 1
There are no transcripts associated with this gene.

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 191 Intron NP_115903.1
NM_181509.2 191 Silent Mutation TCC,TCT S8S NP_852610.1
XM_011529083.2 191 Silent Mutation TCC,TCT S8S XP_011527385.1
XM_011529084.2 191 Silent Mutation TCC,TCT S8S XP_011527386.1
XM_011529085.2 191 Silent Mutation TCC,TCT S8S XP_011527387.1

View Full Product Details