Product Details

SNP ID
rs201405268
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:48625920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCACCGGCGGCTGGCAGAGGC[A/G]GTAGAGGCTGGGGATGGAGGCAAAG
Phenotype
MIM: 606905
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PREX1 PubMed Links

Gene Details

Gene
PREX1
Gene Name
phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020820.3 3870 Missense Mutation CGC,TGC R1649C NP_065871.2
XM_011528934.2 3870 Missense Mutation CGC,TGC R1249C XP_011527236.1

View Full Product Details