Product Details

SNP ID
rs200760388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44697895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGCGGCCGCCTGGCCTCCTGC[A/G]GGTCCCTCCTCTGCCGCCCCACATG
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-12 PubMed Links

Gene Details

Gene
KRTAP10-12
Gene Name
keratin associated protein 10-12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198699.1 724 Missense Mutation AGG,GGG R232G NP_941972.1
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 724 Intron NP_001258966.1
NM_144991.2 724 Intron NP_659428.2

View Full Product Details