Product Details

SNP ID
rs200071559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44540144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCAGACGGACATGGTGGACG[C/T]GGCCATGCTGGGGTGGGGAGGAGGT
Phenotype
MIM: 612920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-1 PubMed Links

Gene Details

Gene
KRTAP10-1
Gene Name
keratin associated protein 10-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198691.2 52 Missense Mutation ACG,GCG T3A NP_941964.2
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 52 Intron NP_001258966.1
NM_144991.2 52 Intron NP_659428.2

View Full Product Details