Product Details
- SNP ID
-
rs201411134
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:25881763 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGCGCTCCTCTGGGGTGACAGC[A/G]GCGTCAACCTGAAAAACAAGAGGAG
- Phenotype
-
MIM: 104760
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41276546] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APP
- Gene Name
- amyloid beta precursor protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000484.3 |
2309 |
Silent Mutation |
GCC,GCT |
A740A |
NP_000475.1 |
NM_001136016.3 |
2309 |
Silent Mutation |
GCC,GCT |
A716A |
NP_001129488.1 |
NM_001136129.2 |
2309 |
Silent Mutation |
GCC,GCT |
A609A |
NP_001129601.1 |
NM_001136130.2 |
2309 |
Silent Mutation |
GCC,GCT |
A684A |
NP_001129602.1 |
NM_001136131.2 |
2309 |
Silent Mutation |
GCC,GCT |
A630A |
NP_001129603.1 |
NM_001204301.1 |
2309 |
Silent Mutation |
GCC,GCT |
A722A |
NP_001191230.1 |
NM_001204302.1 |
2309 |
Silent Mutation |
GCC,GCT |
A703A |
NP_001191231.1 |
NM_001204303.1 |
2309 |
Silent Mutation |
GCC,GCT |
A647A |
NP_001191232.1 |
NM_201413.2 |
2309 |
Silent Mutation |
GCC,GCT |
A721A |
NP_958816.1 |
NM_201414.2 |
2309 |
Silent Mutation |
GCC,GCT |
A665A |
NP_958817.1 |
View Full Product Details