Product Details

SNP ID
rs201202820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231456651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACGGCTTCTGGCATTAGGTGAT[A/C]CCCTGGGTCCTTGCAACTCCAGCCT
Phenotype
MIM: 164035 MIM: 604012 MIM: 611133
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCL PubMed Links

Gene Details

Gene
NCL
Gene Name
nucleolin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005381.2 1826 Missense Mutation GGA,GTA G562V NP_005372.2
Gene
SNORA75
Gene Name
small nucleolar RNA, H/ACA box 75
There are no transcripts associated with this gene.

Gene
SNORD20
Gene Name
small nucleolar RNA, C/D box 20
There are no transcripts associated with this gene.

Gene
SNORD82
Gene Name
small nucleolar RNA, C/D box 82
There are no transcripts associated with this gene.

View Full Product Details