Product Details

SNP ID
rs201575918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27079217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCCAGGCCCAGATTGCCCCCC[A/G]GCCAGCCAGCCGCCACAGGTAAGAG
Phenotype
MIM: 615900 MIM: 130660 MIM: 614058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGBL5 PubMed Links

Gene Details

Gene
AGBL5
Gene Name
ATP/GTP binding protein like 5
There are no transcripts associated with this gene.

Gene
EMILIN1
Gene Name
elastin microfibril interfacer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007046.3 651 Missense Mutation CAG,CGG Q51R NP_008977.1
Gene
KHK
Gene Name
ketohexokinase
There are no transcripts associated with this gene.

Gene
OST4
Gene Name
oligosaccharyltransferase complex subunit 4, non-catalytic
There are no transcripts associated with this gene.

View Full Product Details