Product Details
- SNP ID
-
rs200885816
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:187669196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTCATCCTGCTCAGCAGCCTGGG[A/G]CAGATCTTCAGGTTCCAGGGCATCA
- Phenotype
-
MIM: 182450
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SST
PubMed Links
Gene Details
- Gene
- SST
- Gene Name
- somatostatin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001048.3 |
342 |
Missense Mutation |
CCC,TCC |
P74S |
NP_001039.1 |
View Full Product Details