Product Details
- SNP ID
-
rs200509073
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:31989170 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACTTGACTTTTAGGGATGTGGCT[A/G]TAGAATTCTCTTTGGAGGAGTGGAA
- Phenotype
-
MIM: 606738
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OSBPL10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11129475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OSBPL10
- Gene Name
- oxysterol binding protein like 10
There are no transcripts associated with this gene.
- Gene
- ZNF860
- Gene Name
- zinc finger protein 860
View Full Product Details