Product Details

SNP ID
rs199533328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122893130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACATATCTGACTTCCCAAAAG[C/T]TCCAGGATTTGTGTGCTGTTGCCGA
Phenotype
MIM: 134920 MIM: 606261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF2 PubMed Links

Gene Details

Gene
FGF2
Gene Name
fibroblast growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002006.4 615 UTR 3 NP_001997.5
Gene
NUDT6
Gene Name
nudix hydrolase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007083.4 615 Missense Mutation ACT,GCT T217A NP_009014.2
NM_198041.2 615 Missense Mutation ACT,GCT T48A NP_932158.1

View Full Product Details