Product Details

SNP ID
rs202157766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74933370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGCTCTTTGCATCTTAACTGCA[G/T]GTAATTGGCGCCATCCTCCCGGAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PARM1 PubMed Links

Gene Details

Gene
PARM1
Gene Name
prostate androgen-regulated mucin-like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015393.3 255 Missense Mutation GGA,TGA G15* NP_056208.2
XM_011531833.1 255 Missense Mutation GTT,TTT V15F XP_011530135.1

View Full Product Details