Product Details

SNP ID
rs201201764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144646900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTGCGGTGGCTTCTACCCTCG[G/T]CTGTCCTGCTGCCTGCGGAGTGACA
Phenotype
MIM: 606178
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HHIP PubMed Links

Gene Details

Gene
HHIP
Gene Name
hedgehog interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022475.2 774 Silent Mutation CGG,CGT R75R NP_071920.1
XM_005263178.4 774 Silent Mutation CGG,CGT R75R XP_005263235.1
XM_006714288.3 774 Silent Mutation CGG,CGT R75R XP_006714351.1
Gene
HHIP-AS1
Gene Name
HHIP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details