Product Details

SNP ID
rs201532962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36152268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGGGCCTGCGGACGCTATGCA[C/T]AGGTAAACGGATTGCAAAAAGGGGA
Phenotype
MIM: 601436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMBRD2 PubMed Links

Gene Details

Gene
LMBRD2
Gene Name
LMBR1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007527.1 141 Intron NP_001007528.1
XM_011514162.2 141 Intron XP_011512464.1
XM_017010024.1 141 Intron XP_016865513.1
Gene
MIR580
Gene Name
microRNA 580
There are no transcripts associated with this gene.

Gene
SKP2
Gene Name
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243120.1 141 UTR 5 NP_001230049.1
NM_005983.3 141 Silent Mutation CAC,CAT H2H NP_005974.2
NM_032637.3 141 Silent Mutation CAC,CAT H2H NP_116026.1
XM_011514081.2 141 Silent Mutation CAC,CAT H2H XP_011512383.1
XM_011514082.2 141 Silent Mutation CAC,CAT H2H XP_011512384.1
XM_011514083.2 141 UTR 5 XP_011512385.1
XM_017009753.1 141 UTR 5 XP_016865242.1

View Full Product Details