Product Details

SNP ID
rs201338573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:476217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTTACCCGCTTCTGGGCACG[C/T]TCCCGCTTGTACAGCTTGGCCGCCT
Phenotype
MIM: 608186 MIM: 182307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC3 PubMed Links

Gene Details

Gene
EXOC3
Gene Name
exocyst complex component 3
There are no transcripts associated with this gene.

Gene
LOC100288152
Gene Name
uncharacterized LOC100288152
There are no transcripts associated with this gene.

Gene
PP7080
Gene Name
uncharacterized LOC25845
There are no transcripts associated with this gene.

Gene
SLC9A3
Gene Name
solute carrier family 9 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284351.2 2164 Silent Mutation NP_001271280.1
NM_004174.3 2164 Silent Mutation NP_004165.2

View Full Product Details