Product Details

SNP ID
rs201829505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33398291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTCTTTATCTTTCCCCAGAAA[C/T]GGACAAGAGGCCTGGGTGCAACGAC
Phenotype
MIM: 603763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIFC1 PubMed Links

Gene Details

Gene
KIFC1
Gene Name
kinesin family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002263.3 310 Missense Mutation CGG,TGG R52W NP_002254.2
XM_011514585.1 310 Missense Mutation CGG,TGG R52W XP_011512887.1
XM_011514587.2 310 Missense Mutation CGG,TGG R52W XP_011512889.1
XM_017010836.1 310 Missense Mutation CGG,TGG R52W XP_016866325.1
XM_017010837.1 310 Missense Mutation CGG,TGG R11W XP_016866326.1

View Full Product Details