Product Details
- SNP ID
-
rs201957387
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26216368 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGCACGGCGGTCTGGATCTCCCT[A/G]GAGGTAATGGTCGAGCGCTTGTTGT
- Phenotype
-
MIM: 602786
MIM: 602798
MIM: 602813
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H2AE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16891427] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H2AE
- Gene Name
- histone cluster 1, H2ae
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021052.2 |
|
Intron |
|
|
NP_066390.1 |
- Gene
- HIST1H2BG
- Gene Name
- histone cluster 1, H2bg
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003518.3 |
|
Intron |
|
|
NP_003509.1 |
- Gene
- HIST1H3E
- Gene Name
- histone cluster 1, H3e
There are no transcripts associated with this gene.
View Full Product Details