Product Details

SNP ID
rs199672479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127578734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAAACTGTGGCTATGAATCTG[C/G]AGATCATATGTGCATCTCTGTGGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf58 PubMed Links

Gene Details

Gene
C6orf58
Gene Name
chromosome 6 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010905.2 362 Missense Mutation GCA,GGA A117G NP_001010905.1

View Full Product Details