Product Details

SNP ID
rs200999878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22727598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACGAATTGACAAACAAATTCG[A/G]TACATCCTCGACGGCATCTCAGCCC
Phenotype
MIM: 147620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL6 PubMed Links

Gene Details

Gene
IL6
Gene Name
interleukin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000600.4 628 Silent Mutation CGA,CGG R58R NP_000591.1
NM_001318095.1 628 Intron NP_001305024.1
XM_005249745.4 628 Silent Mutation CGA,CGG R112R XP_005249802.1
XM_011515390.2 628 Silent Mutation CGA,CGG R58R XP_011513692.1
Gene
LOC541472
Gene Name
uncharacterized LOC541472
There are no transcripts associated with this gene.

View Full Product Details