Product Details

SNP ID
rs200321023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66633323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGGAGGGGCTCACTTCACTACA[C/T]GCCTGTCCACACTGCGGTGCTACGA
Phenotype
MIM: 611725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD7 PubMed Links

Gene Details

Gene
KCTD7
Gene Name
potassium channel tetramerization domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167961.2 377 Missense Mutation CGC,TGC R65C NP_001161433.1
NM_153033.4 377 Missense Mutation CGC,TGC R65C NP_694578.1

View Full Product Details