Product Details
- SNP ID
-
rs200213430
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151381935 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAACACCCTCCGCAGCTCTCCAGA[C/T]TGCGCGTCGAAGGCCCGGGCGCAAG
- Phenotype
-
MIM: 607981
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUB1
PubMed Links
Gene Details
- Gene
- NUB1
- Gene Name
- negative regulator of ubiquitin like proteins 1
There are no transcripts associated with this gene.
- Gene
- WDR86
- Gene Name
- WD repeat domain 86
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284260.1 |
1355 |
Missense Mutation |
ATC,GTC |
I325V |
NP_001271189.1 |
NM_001284261.1 |
1355 |
Missense Mutation |
AAT,AGT |
N130S |
NP_001271190.1 |
NM_001284262.1 |
1355 |
Silent Mutation |
CAA,CAG |
Q175Q |
NP_001271191.1 |
NM_198285.2 |
1355 |
Silent Mutation |
CAA,CAG |
Q303Q |
NP_938026.2 |
XM_005249989.3 |
1355 |
Missense Mutation |
AAT,AGT |
N258S |
XP_005250046.1 |
XM_005249990.4 |
1355 |
Silent Mutation |
CAA,CAG |
Q303Q |
XP_005250047.1 |
XM_006715966.2 |
1355 |
Intron |
|
|
XP_006716029.1 |
XM_011516144.2 |
1355 |
Missense Mutation |
ATC,GTC |
I325V |
XP_011514446.1 |
XM_011516145.2 |
1355 |
Missense Mutation |
ATC,GTC |
I325V |
XP_011514447.1 |
XM_011516146.2 |
1355 |
Silent Mutation |
CAA,CAG |
Q303Q |
XP_011514448.1 |
XM_011516147.2 |
1355 |
Missense Mutation |
AAT,AGT |
N258S |
XP_011514449.1 |
XM_011516148.1 |
1355 |
Missense Mutation |
ATC,GTC |
I197V |
XP_011514450.1 |
XM_011516150.1 |
1355 |
Missense Mutation |
ATC,GTC |
I197V |
XP_011514452.1 |
XM_011516151.2 |
1355 |
Missense Mutation |
AAT,AGT |
N258S |
XP_011514453.1 |
XM_011516152.1 |
1355 |
Intron |
|
|
XP_011514454.1 |
View Full Product Details