Product Details

SNP ID
rs200560520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144004483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCCCTGGCGAAGATCTACTTC[A/C]TCTCCTGCCTCAGCTTCTGTGGTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR6B1 PubMed Links

Gene Details

Gene
OR6B1
Gene Name
olfactory receptor family 6 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005281.1 487 Missense Mutation ATC,CTC I163L NP_001005281.1

View Full Product Details